About this item
HYDRA-POINT® coulometric reagents are designed to deliver faster analyses, lower detection limits and increased accuracy.
- Pyridine- and chlorocarbon-free vessel solution
- The fritless/diaphragmless reagent works both in cells without and with diaphragm
- High water capacity, where the anolyte can take over 1000 mg of water per charge
These products feature lower background noise that gives a better signal/noise ratio, resulting in a significant increase in the lower detection limit—to as low as 1 ppm.