About this item
Sklar's® Titanium Micro Dressing Forceps are a lighter version of their stainless steel counterparts.
- Non-magnetic
- Lighter than stainless steel
- Durable and corrosion resistant
- Reusable and nonsterile
The forceps offer superior durability and corrosion resistance. Because they are non-magnetic, they are also MRI compatible.