Ward's® Tryptic Soy Broth Dehydrated Media, Powdered, IPM Scientific
Catalog #: 470024-770
Supplier: IPM SCIENTIFIC, INC.
CAS Number:
Specifications
- Packaging:Bottle
- Description:Tryptic Soy Broth
- Size:500 g
- Cat. no.:470024-770
- Supplier No.:M011-500G
Specifications
About this item
Dilute quantities to any volume as needed.
- Economical alternative
- Stores much longer than prepared media
- Prepare only as much as you need at one time
Use Ward's dehydrated media for many microbiology applications. Preparation instructions are included.
Customers Who Bought This Also Bought