Ward's® Sabouraud Dextrose Agar Dehydrated Media, Powdered, IPM Scientific
Catalog #: 470024-758
Supplier: IPM SCIENTIFIC, INC.
CAS Number:
Specifications
- Packaging:Bottle
- Description:Sabouraud Dextrose Agar
- Size:500 g
- Cat. no.:470024-758
- Supplier No.:381006
Specifications
About this item
Dilute quantities to any volume as needed.
- Economical alternative
- Stores much longer than prepared media
- Prepare only as much as you need at one time
- 65g/L
Use Ward's dehydrated media for many microbiology applications. Preparation instructions are included.
Customers Who Bought This Also Bought