Syringe Adaptor Kit for Single-Stage VOC Regulator, Restek
Catalog #: 10854-536
Supplier: Restek
Specifications
- Description:Syringe Adaptor Kit
- Cat. no.:10854-536
- Supplier No.:21118
Specifications
About this item
Use to withdraw sample from a high-pressure cylinder after pressure reduction through the high-purity VOC single-stage regulator.
- Withdraw sample after pressure reduction
- High-purity regulator
- Use with any syringe needle
Kit contains one nickel-plated brass 1/4" NPT to female luer fitting, which can be used with an A-2 Luer syringe, and one stainless steel 1/4" NPT×1/8" compression fitting with septum (can be used with any syringe needle).
Customers Who Bought This Also Bought