Deuterium Lamp for Agilent HPLC Systems, Restek
Catalog #: 10830-256
Supplier: Restek
Specifications
- Description:Deuterium Lamp for Agilent 1100 DAD/MWD/1200 (Compatible with 1260)
- Cat. no.:10830-256
- Bulb Life:2000 h
- Supplier No.:25888
Specifications
About this item
Deuterium lamp, 1100 DAD/MWD/1200, for HPLC systems has life span of 2000 hours.
Customers Who Bought This Also Bought