Coping Saw
Catalog #: 470228-938
Supplier: KIDDER MFG. CO. LTD
Specifications
- Description:Coping Saw
- Cat. no.:470228-938
Specifications
About this item
Free style cutting tool.
- The blade has 5 points
This professional quality coping saw has a sturdy steel frame. Blade tension is achieved by turning the wood handle. Easily adjustable blade fixtures and an extra narrow specially hardened blade are among the features.
Customers Who Bought This Also Bought