BioClean™ Sterile Anti-Fog Spray, Ansell
Catalog # 75972-514
Supplier: Ansell Healthcare
Specifications
- Description:BioClean™ Sterile Anti-Fog Solution for Autoclavable Goggles
- Size:60 ml
- Environmentally Preferable:
- Cat. no.:75972-514
- Supplier No.:BAFS-601
Specifications
About this item
Spray cleaner for cleanroom eyewear, compatible with all BioClean Clearview™ goggles.
- Eliminates fogging
- Quick and easy application
- Alcohol-free
Packaging: One bottle per inner bag, 10 bags per outer bag, 10 bags (100 bottles) per carton.