Column Cutters, PerkinElmer
Catalog #: CA97019-380
Supplier: PERKINELMER SCIENTIFIC CANADA ULC
Specifications
- Description:Column Cutter
- Cat. no.:CA97019-380
- Supplier No.:N9301376
Specifications
About this item
This ceramic column cutter is ideal for cutting polyimide fused silica capillary columns and guard columns.
- Easy to hold and simple to use
- All four sides can be used as a cutting tool
Customers Who Bought This Also Bought