To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:Neat
- Description:Fatty acid methyl ester, FAME Std #7
- Size:50 mg
- Cat. no.:RK35016
- Supplier No.:35016
Specifications
About this item
Contains: Chain, Compound (CAS#), % by Weight; C6:00 Methyl caproate (106-70-7), 20%; C7:00 Methyl heptanoate (106-73-0), 20%; C8:00 Methyl caprylate (111-11-5), 20%; C9:00 Methyl nonanoate (1731-84-6), 20%; C10:00 Methyl caprate (110-42-9), 20%