Order Entry
ContactUsLinkComponent
Herbicide Lab Performance Check Mix (5 components), Restek
Herbicide Lab Performance Check Mix (5 components), Restek
Catalog #: RK32063
Supplier:  Restek
CAS Number:  
Herbicide Lab Performance Check Mix for EPA 515
Herbicide Lab Performance Check Mix (5 components), Restek
Catalog #: RK32063
Supplier:  Restek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    In methyl tert-butyl ether
  • Description:
    Herbicide Lab Performance Check Mix
  • Size:
    1 ml/Ampule
  • Cat. no.:
    RK32063
  • Certification:
    CRM
  • Supplier No.:
    32063

Specifications

About this item

This mix contains five components.

Components are dinoseb methyl ether (4 μg/ml), DCAA methyl ester (500 μg/ml), 4,4'-dibromooctafluorobiphenyl (250 μg/ml), 3,5-dichlorobenzoic acid methyl ester (600 µg/ml), 4-nitroanisole (1600 μg/ml).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Customers Who Bought This Also Bought