To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:1000 µg/ml each in methylene chloride (3-methylphenol and 4-methylphenol at 500 µg/ml)
- Description:8270 MegaMix
- Size:1 ml/Ampule
- Cat. no.:RK31850
- Certification:CRM
- Supplier No.:31850
Specifications
About this item
Contains: Acenaphthene (83-32-9); Acenaphthylene (208-96-8); Aniline (62-53-3); Anthracene (120-12-7); Azobenzene (103-33-3)*; Benz(a)anthracene (56-55-3); Benzo(a)pyrene (50-32-8); Benzo(b)fluoranthene (205-99-2); Benzo(ghi)perylene (191-24-2); Benzo(k)fluoranthene (207-08-9); Benzyl alcohol (100-51-6); Benzyl butyl phthalate (85-68-7); Bis(2-chloroethoxy)methane (111-91-1); Bis(2-chloroethyl)ether (111-44-4); Bis(2-ethylhexyl)adipate (103-23-1); Bis(2-ethylhexyl)phthalate (117-81-7); 4-Bromophenyl phenyl ether (101-55-3); Carbazole (86-74-8); 4-Chloroaniline (106-47-8); 4-Chloro-3-methylphenol (59-50-7); 2-Chloronaphthalene (91-58-7); 2-Chlorophenol (95-57-8); 4-Chlorophenyl phenyl ether (7005-72-3); Chrysene (218-01-9); Dibenz(a,h)anthracene (53-70-3); Dibenzofuran (132-64-9); 1,2-Dichlorobenzene (95-50-1); 1,3-Dichlorobenzene (541-73-1); 1,4-Dichlorobenzene (106-46-7); 2,4-Dichlorophenol (120-83-2); Diethylphthalate (84-66-2); 2,4-Dimethylphenol (105-67-9); Dimethylphthalate (131-11-3); Di-n-butyl phthalate (84-74-2); 1,2-Dinitrobenzene (528-29-0); 1,3-Dinitrobenzene (99-65-0); 1,4-Dinitrobenzene (100-25-4); 4,6-Dinitro-2-methylphenol (Dinitro-o-cresol) (534-52-1); 2,4-Dinitrophenol (51-28-5); 2,4-Dinitrotoluene (121-14-2); 2,6-Dinitrotoluene (606-20-2); Di-n-octyl phthalate (117-84-0); Diphenylamine (122-39-4)**; Fluoranthene (206-44-0); Fluorene (86-73-7); Hexachlorobenzene (118-74-1); Hexachlorobutadiene (87-68-3); Hexachlorocyclopentadiene (77-47-4); Hexachloroethane (67-72-1); Indeno(1,2,3-cd)pyrene (193-39-5); Isophorone (78-59-1); 1-Methylnaphthalene (90-12-0); 2-Methylnaphthalene (91-57-6); 2-Methylphenol (o-cresol) (95-48-7); 3-Methylphenol (m-cresol) (108-39-4); 4-Methylphenol (p-cresol) (106-44-5); Naphthalene (91-20-3); 2-Nitroaniline (88-74-4); 3-Nitroaniline (99-09-2); 4-Nitroaniline (100-01-6); Nitrobenzene (98-95-3); 2-Nitrophenol (88-75-5); 4-Nitrophenol (100-02-7); N-Nitrosodimethylamine (62-75-9); N-Nitroso-di-n-propylamine (621-64-7); 2,2'-Oxybis(1-chloropropane) (108-60-1); Pentachlorophenol (87-86-5); Phenanthrene (85-01-8); Phenol (108-95-2); Pyrene (129-00-0); Pyridine (110-86-1); 2,3,4,6-Tetrachlorophenol (58-90-2); 2,3,5,6-Tetrachlorophenol (935-95-5); 1,2,4-Trichlorobenzene (120-82-1); 2,4,5-Trichlorophenol (95-95-4); 2,4,6-Trichlorophenol (88-06-2)
- Fewest mixtures needed for calibration and matrix spikes.
- Mixtures formulated for maximum stability.
- Contains most routinely analyzed compounds.