Specifications
- Concentration:50000 µg/ml in methylene chloride
- Description:Used motor oil composite
- Size:1 ml/Ampule
- Cat. no.:RK31465
- Certification:CRM
- Supplier No.:31465
Specifications
About this item
Prepared from an equal-volume blend from five gasoline-powered vehicles. After blending, a precisely weighed amount of the composite is added to a volumetric flask to produce the standard. Motor Oil standards may exhibit lot-to-lot variation.