Florida TRPH Standard (17 components), Restek
Catalog #: RK31266
Supplier: Restek
CAS Number:
Specifications
- Concentration:500 µg/ml each in hexane
- Description:Florida TRPH Standard
- Size:1 ml/Ampule
- Cat. no.:RK31266
- Certification:CRM
- Supplier No.:31266
Specifications
About this item
This contains n-octane (C8), n-decane (C10), n-dodecane (C12), n-tetradecane (C14), n-hexadecane (C16), n-octadecane (C18), n-eicosane (C20), n-docosane (C22), n-tetracosane (C24), n-hexacosane (C26), n-octacosane (C28), n-triacontane (C30), n-dotriacontane (C32), n-tetratriacontane (C34), n-hexatriacontane (C36), n-octatriacontane (C38) and n-tetracontane (C40).
Customers Who Bought This Also Bought