To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:2000 µg/ml each in methylene chloride
- Description:Fuel Oil Degradation Mix
- Size:1 ml/Ampule
- Cat. no.:RK31240
- Certification:CRM
- Supplier No.:31240
Specifications
About this item
Subsurface degradation of fuel oil spills can be estimated by examining the ratios of C17/pristane and C18/phytane. To assist in identifying these four compounds from the complex fuel oil analysis, Restek offer a product that contains these compounds for retention time determination.
Contains: (C17) Heptadecane (629-78-7); (C18) Octadecane (593-45-3); Pristane (2,6,10,14-tetramethylpentadecane) (1921-70-6); Phytane (2,6,10,14-tetramethylhexadecane) (638-36-8)