To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:D3710-95 Calibration Mix
- Size:1 ml/Ampule
- Cat. no.:RK31223
- Certification:CRM
- Supplier No.:31223
Specifications
About this item
These calibration mixtures are made with pure, highly characterized neat material and are prepared using NIST-traceable balances and weights. Each ampul is supplied with a data sheet indicating the exact concentration and a sample chromatogram.
Contains: Compound (CAS#), Conc. (% vol/vol); (C5) n-Pentane (109-66-0), 8%; (C6) n-Hexane (110-54-3), 6%; (C7) n-Heptane (142-82-5), 10%; (C8) n-Octane (111-65-9), 5%; (C10) n-Decane (124-18-5), 4%; (C12) n-Dodecane (112-40-3), 4%; (C13) n-Tridecane (629-50-5), 2%; (C14) n-Tetradecane (629-59-4), 2%; (C15) n-Pentadecane (629-62-9), 2%; n-Butylbenzene (104-51-8), 4%; 2,4-Dimethylpentane (108-08-7), 6%; 2-Methylbutane (Isopentane) (78-78-4), 10%; 2-Methylpentane (107-83-5), 6%; n-Propylbenzene (103-65-1), 5%; Toluene (108-88-3), 12%; p-Xylene (106-42-3), 14%