To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:1000 µg/ml each in methylene chloride
- Description:DRO Mix (Tennessee/Mississippi)
- Size:1 ml/Ampule
- Cat. no.:RK31214
- Certification:CRM
- Supplier No.:31214
Specifications
About this item
Contains: (C10) n-Decane (124-18-5); (C11) n-Undecane (1120-21-4); (C12) n-Dodecane (112-40-3); (C13) n-Tridecane (629-50-5); (C14) n-Tetradecane (629-59-4); (C15) n-Pentadecane (629-62-9); (C16) n-Hexadecane (544-76-3); (C17) n-Heptadecane (629-78-7); (C18) n-Octadecane (593-45-3); (C19) n-Nonadecane (629-92-5); (C20) n-Eicosane (112-95-8); (C21) n-Heneicosane (629-94-7); (C22) n-Docosane (629-97-0); (C23) n-Tricosane (638-67-5); (C24) n-Tetracosane (646-31-1); (C25) n-Pentacosane (629-99-2)