To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:10000 µg/ml in P and T methanol
- Description:Stoddard solvent
- Size:1 ml/Ampule
- Cat. no.:RK30487
- Certification:CRM
- Supplier No.:30487
Specifications
About this item
Weathered Petrochemical Standards
Stoddard solvent is also known as Type I mineral spirits, Texsolve S, or Varsol 1 mineral spirits. We offer this reference material for those who need to calibrate Stoddard solvent separately. This standard is dissolved in methanol for analysis by either direct injection or purge-and-trap.
These solutions are prepared from a single-source (one-refinery) product. The weathered materials indicate the percent weight loss from the original material. Samples of regular- and premium-grade unleaded gasoline were collected, then blended in equal volumes.There are four general types of mineral spirits, classified according to boiling point range (BPR):
Type I (Stoddard solvent), BPR 149 to 182 °C
Type II (high flash point), BPR 177 to 196 °C
Type III (odorless), BPR 149 to 196 °C
Type IV (low dry point), BPR 149 to 174 °C
Stoddard solvent, a petroleum distillate mixture of C7-C12 hydrocarbons, consists of three major groups of components: linear and branched alkanes-also known as paraffins (30 to 50%); cycloalkanes-also called cycloparaffins or naphthenes (30 to 40%); and aromatic hydrocarbons (10 to 20%).
Contains: Stoddard solvent (8052-41-3)