To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:Contains 1 ml each of these mixtures
- Description:524 Rev. 4.0 VOA Kit #2, 2,000ug/mL
- Size:Kit
- Cat. no.:RK30447
- Certification:CRM
- Supplier No.:30447
Specifications
About this item
Contains: 30042: 502.2 Calibration Mix #1 (gases); Bromomethane (methyl bromide) (74-83-9); Chloroethane (ethyl chloride) (75-00-3); Chloromethane (methyl chloride) (74-87-3); Dichlorodifluoromethane (CFC-12) (75-71-8); Trichlorofluoromethane (CFC-11) (75-69-4); Vinyl chloride (75-01-4);30431: 502.2 MegaMix Standard; Benzene (71-43-2); Bromobenzene (108-86-1); Bromochloromethane (74-97-5); Bromodichloromethane (75-27-4); Bromoform (75-25-2); n-Butylbenzene (104-51-8); sec-Butylbenzene (135-98-8); tert-Butylbenzene (98-06-6); Carbon tetrachloride (56-23-5); Chlorobenzene (108-90-7); Chloroform (67-66-3); 2-Chlorotoluene (95-49-8); 4-Chlorotoluene (106-43-4); Dibromochloromethane (124-48-1); 1,2-Dibromo-3-chloropropane (DBCP) (96-12-8); 1,2-Dibromoethane (EDB) (106-93-4); Dibromomethane (74-95-3); 1,2-Dichlorobenzene (95-50-1); 1,3-Dichlorobenzene (541-73-1); 1,4-Dichlorobenzene (106-46-7); 1,1-Dichloroethane (75-34-3); 1,2-Dichloroethane (107-06-2); 1,1-Dichloroethene (75-35-4); cis-1,2-Dichloroethene (156-59-2); trans-1,2-Dichloroethene (156-60-5); 1,2-Dichloropropane (78-87-5); 1,3-Dichloropropane (142-28-9); 2,2-Dichloropropane (594-20-7); 1,1-Dichloropropene (563-58-6); cis-1,3-Dichloropropene (10061-01-5); trans-1,3-Dichloropropene (10061-02-6); Ethylbenzene (100-41-4); Hexachloro-1,3-butadiene (hexachlorobutadiene) (87-68-3); Isopropylbenzene (cumene) (98-82-8); 4-Isopropyltoluene (p-cymene) (99-87-6); Methylene chloride (dichloromethane) (75-09-2); Naphthalene (91-20-3); n-Propylbenzene (103-65-1); Styrene (100-42-5); 1,1,1,2-Tetrachloroethane (630-20-6); 1,1,2,2-Tetrachloroethane (79-34-5); Tetrachloroethene (127-18-4); Toluene (108-88-3); 1,2,3-Trichlorobenzene (87-61-6); 1,2,4-Trichlorobenzene (120-82-1); 1,1,1-Trichloroethane (71-55-6); 1,1,2-Trichloroethane (79-00-5); Trichloroethene (79-01-6); 1,2,3-Trichloropropane (96-18-4); 1,2,4-Trimethylbenzene (95-63-6); 1,3,5-Trimethylbenzene (108-67-8); m-Xylene (108-38-3); o-Xylene (95-47-6); p-Xylene (106-42-3);30300: 524 Calibration Mix #7A; Acetone (67-64-1); 2-Butanone (MEK) (78-93-3); 2-Hexanone (591-78-6); 4-Methyl-2-pentanone (MIBK) (108-10-1); Tetrahydrofuran (109-99-9);30304: 524 Calibration Mix #7B; Acrylonitrile (107-13-1); Allyl chloride (3-chloropropene) (107-05-1); Ethyl methacrylate (97-63-2); Methyl acrylate (96-33-3); Methyl methacrylate (80-62-6); Nitrobenzene (98-95-3); Pentachloroethane (76-01-7);30203: 524 Calibration Mix #8; Carbon disulfide (75-15-0); Chloroacetonitrile (107-14-2); 1-Chlorobutane (Butyl chloride) (109-69-3); trans-1,4-Dichloro-2-butene (110-57-6); 1,1-Dichloro-2-propanone (513-88-2); Diethyl ether (ethyl ether) (60-29-7); Hexachloroethane (67-72-1); Iodomethane (methyl iodide) (74-88-4); Methacrylonitrile (126-98-7); Methyl tert-butyl ether (MTBE) (1634-04-4); 2-Nitropropane (79-46-9); Propionitrile (107-12-0);30201: 524 Internal Standard/Surrogate Mix; 1-Bromo-4-fluorobenzene (BFB) (460-00-4); 1,2-Dichlorobenzene-d4 (2199-69-1); Fluorobenzene (462-06-6)