To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:Contains 1 ml each of these mixtures
- Description:Weathered Gasoline Kit #2
- Size:Kit
- Cat. no.:RK30437
- Certification:CRM
- Supplier No.:30437
Specifications
About this item
These solutions are prepared from a single-source (one-refinery) product. The weathered materials indicate the percent weight loss from the original material. Samples of regular- and premium-grade unleaded gasoline were collected, then blended in equal volumes.
Contains: 30096: Unleaded Gasoline (ASTM Class 2 Accelerant); Unleaded gasoline: unweathered (8006-61-9);30097: Unleaded Gasoline (ASTM Class 2 Accelerant); Unleaded gasoline: 25% weathered (8006-61-9);30098: Unleaded Gasoline (ASTM Class 2 Accelerant); Unleaded gasoline: 50% weathered (8006-61-9);30099: Unleaded Gasoline (ASTM Class 2 Accelerant); Unleaded gasoline: 75% weathered (8006-61-9);30436: Unleaded Gasoline (ASTM Class 2 Accelerant); Unleaded gasoline: 99% weathered (8006-61-9)