To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:2000 µg/ml each in P and T methanol
- Description:PA DEP UST Standard
- Size:1 ml/Ampule
- Cat. no.:RK30433
- Certification:CRM
- Supplier No.:30433
Specifications
About this item
Contains: Benzene (71-43-2); 1,2-Dibromoethane (EDB) (106-93-4); 1,2-Dichloroethane (107-06-2); Ethylbenzene (100-41-4); Isopropyl benzene (cumene) (98-82-8); Methyl tert-butyl ether (MTBE) (1634-04-4); Naphthalene (91-20-3); Toluene (108-88-3); m-Xylene (108-38-3); o-Xylene (95-47-6); p-Xylene (106-42-3)