To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:Contains 1 ml each of these mixtures
- Description:DW-VOC Kit #2
- Size:Kit
- Cat. no.:RK30221
- Certification:CRM
- Supplier No.:30221
Specifications
About this item
Contains: 30211: 501 Trihalomethane Mix; Bromodichloromethane (75-27-4); Bromoform (75-25-2); Chloroform (67-66-3); Dibromochloromethane (124-48-1);30219: DW-VOC Mix #1; Benzene (71-43-2); Carbon tetrachloride (56-23-5); 1,4-Dichlorobenzene (106-46-7); 1,2-Dichloroethane (107-06-2); 1,1-Dichloroethene (75-35-4); 1,1,1-Trichloroethane (71-55-6); Trichloroethene (79-01-6); Vinyl chloride (75-01-4);30220: DW-VOC Mix #2; Chlorobenzene (108-90-7); 1,2-Dichlorobenzene (95-50-1); cis-1,2-Dichloroethene (156-59-2); trans-1,2-Dichloroethene (156-60-5); 1,2-Dichloropropane (78-87-5); Ethylbenzene (100-41-4); Styrene (100-42-5); Tetrachloroethene (127-18-4); Toluene (108-88-3); m-Xylene (108-38-3); o-Xylene (95-47-6); p-Xylene (106-42-3);30235: DW-VOC Mix #3; Methylene chloride (dichloromethane) (75-09-2); 1,2,4-Trichlorobenzene (120-82-1); 1,1,2-Trichloroethane (79-00-5)