Unleaded Gasoline Composite Standard, Restek
Supplier: Restek
Certificates
About this item
Contains: Unleaded gasoline composite (8006-61-9)
- Unleaded gasoline composite standards are comprised of three separate sources each of 87, 89, and 93 octane blended in equal portions.
- Gasoline standards may exhibit lot-to-lot variation.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.