Specifications
- Concentration:2000 µg/ml each in P and T methanol
- Description:524 Calibration Mix #8
- Size:1 ml/Ampule
- Cat. no.:RK30203
- Certification:CRM
- Supplier No.:30203
Specifications
About this item
Mix Contains: Carbon disulfide (75-15-0); Chloroacetonitrile (107-14-2); 1-Chlorobutane (Butyl chloride) (109-69-3); trans-1,4-Dichloro-2-butene (110-57-6); 1,1-Dichloro-2-propanone (513-88-2); Diethyl ether (ethyl ether) (60-29-7); Hexachloroethane (67-72-1); Iodomethane (methyl iodide) (74-88-4); Methacrylonitrile (126-98-7); Methyl tert-butyl ether (MTBE) (1634-04-4); 2-Nitropropane (79-46-9); Propionitrile (107-12-0).