8270 Calibration Mix #1 (19 components), Restek
Catalog #: 10118-478
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml each in methylene chloride
- Description:8270 Calibration Mix #1
- Size:1 ml/Ampule
- Cat. no.:10118-478
- Certification:CRM
- Supplier No.:31618
Specifications
About this item
Contains: Benzoic acid (65-85-0); 4-Chloro-3-methylphenol (59-50-7); 2-Chlorophenol (95-57-8); 2,4-Dichlorophenol (120-83-2); 2,6-Dichlorophenol (87-65-0); 2,4-Dimethylphenol (105-67-9); 4,6-Dinitro-2-methylphenol (Dinitro-o-cresol) (534-52-1); 2,4-Dinitrophenol (51-28-5); Dinoseb (88-85-7); 2-Methylphenol (o-cresol) (95-48-7); 3-Methylphenol (m-cresol) (108-39-4); 4-Methylphenol (p-cresol) (106-44-5); 2-Nitrophenol (88-75-5); 4-Nitrophenol (100-02-7); Pentachlorophenol (87-86-5); Phenol (108-95-2); 2,3,4,6-Tetrachlorophenol (58-90-2); 2,4,5-Trichlorophenol (95-95-4); 2,4,6-Trichlorophenol (88-06-2)
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Customers Who Bought This Also Bought