Fortification Mix (7 components), Restek
Catalog #: 10118-436
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml each in methylene chloride
- Description:Fortification Mix
- Size:1 ml/Ampule
- Cat. no.:10118-436
- Certification:CRM
- Supplier No.:31813
Specifications
About this item
Contains: 4,6-Dinitro-2-methylphenol (Dinitro-o-cresol) (534-52-1); 2,4-Dinitrophenol (51-28-5); 2-Nitroaniline (88-74-4); 3-Nitroaniline (99-09-2); 4-Nitroaniline (100-01-6); 4-Nitrophenol (100-02-7); 2,4,5-Trichlorophenol (95-95-4)
Customers Who Bought This Also Bought