XERT-500 Column Test Mix (8 Components), Restek
Catalog # 10058-066
Supplier: Restek
CAS Number:
Specifications
- Concentration:500 µg/ml each in toluene
- Description:XERT-500 Column Test Mix
- Size:1 ml/Ampule
- Cat. no.:10058-066
- Certification:CRM
- Supplier No.:35247
Specifications
About this item
This contains (C9:0 FAME) methyl nonanoate, (C13) n-tridecane, (C15) n-pentadecane, acenaphthylene, 4-chlorophenol, n-decylamine, 1,6-hexanediol and 1-undecanol (112-42-5).
Frequently Bought Together