NPI-350 Column Test Mix (8 components), Restek
Catalog #: 10058-064
Supplier: Restek
CAS Number:
Specifications
- Concentration:350 µg/ml each in methylene chloride
- Description:NPI-350 Column Test Mix
- Size:1 ml/Ampule
- Cat. no.:10058-064
- Certification:CRM
- Supplier No.:35225
Specifications
About this item
Contains: (C9:0 FAME) Methyl nonanoate (1731-84-6); (C13) n-Tridecane (629-50-5); (C15) n-Pentadecane (629-62-9); Acenaphthylene (208-96-8); 4-Chlorophenol (106-48-9); 1,6-Hexanediol (629-11-8); 4-Propylaniline (2696-84-6); 1-Undecanol (112-42-5)
Customers Who Bought This Also Bought