Labeled Pesticide Residue Internal Standards for Food Analysis, Restek
Supplier: Restek
Certificates
About this item
- Isotopically labeled to provide the best approach for pesticide residue quantification
- Multiple options let you choose internal and surrogate standards that will mitigate matrix effects
- Economically priced and compatible with both LC-MS and GC-MS applications; even helpful for optimizing LC-MS/MS system performance
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.