SP Wilmad-LabGlass Chromatography Columns with Fritted Disc, SP Industries
Supplier: Avantor
Certificates
About this item
These borosilicate glass plain chromatography columns have a coarse porosity fritted disc sealed into the column just above the straight bore stopcock with PTFE plug.
Customers Who Bought This Also Bought