Grinding Balls, U.S. Stoneware
Catalog #: 48906-647
Supplier: US Stoneware
Specifications
- Ø:12.7 mm
- Description:Grinding Balls
- Material:Aluminium
- Cat. no.:48906-647
- Supplier No.:HDAL050
Specifications
About this item
Grinding balls are made of high density aluminium to use with grinding mills.
Customers Who Bought This Also Bought