ContactUsLinkComponent
SolderMag Magnifier
SolderMag Magnifier
Catalog #: 470227-878
Supplier:  CARSON OPTICAL
SolderMag* solder station magnifier
SolderMag Magnifier
Catalog #: 470227-878
Supplier:  CARSON OPTICAL
Specifications
  • Magnification:
    1.75×, 4.5×
  • Description:
    SolderMag magnifier
  • Cat. no.:
    470227-878
Specifications
About this item
A 1.75X hands free solder station magnifier with LED's.
  • 4.5X spot lens
  • Soldering holder
  • Solder spool dispenser
  • 16 LED's
  • Use a standard AC/DC outlet or 4 AA batteries
Customers Who Bought This Also Bought