Order Entry
ContactUsLinkComponent
Pierce PeroXOquant™ Quantitative Peroxide Assay Kits (Lipid), Thermo Scientific
Pierce PeroXOquant™ Quantitative Peroxide Assay Kits (Lipid), Thermo Scientific
Catalog #: CAPI23285
Supplier:  Thermo Scientific
CAS Number:  
Quantitative Peroxide Assay Kit (Lipid)
Pierce PeroXOquant™ Quantitative Peroxide Assay Kits (Lipid), Thermo Scientific
Catalog #: CAPI23285
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Lipid Compatible Formulation Kit
  • Cat. no.:
    CAPI23285
  • No. of Assays/Kit:
    100 tube assays or 500 microplate assays with lipid-containing samples
  • Supplier No.:
    23285

Specifications

About this item

Pierce™ Quantitative Peroxide Assay Kit (Lipid) detects and measures hydrogen peroxide levels in biological samples using an Iron and Xylenol orange* (XO) reagent.

  • Ready to use – removes the variability, difficulty and expense of formulating in-house
  • Ideal for sensitive measurement of peroxide accumulation in non-lipid samples

This Lipid-compatible peroxide assay kit detects peroxides based on oxidation of ferrous to ferric ion in the presence of xylenol orange

Peroxides in the sample oxidize Fe2+ to Fe3+. The Fe3+ ion will form a colored complex with xylenol orange that can be read at 560nm.

*Xylenol orange is (o-cresolsulfonephthalein-3´-3´-bis-[methyliminodiacetic acid sodium salt]).

It can be used with lipid-containing samples without extracting the lipids. The assay is less sensitive, since it does not contain sorbitol. Most proteins do not interfere with this assay, although some metal chelators may require use of a blank (ie. control) to mitigate these effects.

Delivery: Complete kit includes Reagent A (1 mL) and Reagent C (4x25 mL).
Customers Who Bought This Also Bought