SP Wilmad-LabGlass Lattice Clamps, SP Industries
Supplier: SP Industries
Certificates
About this item
These stamped steel clamps are zinc plated for corrosion resistance and have a steel thumbscrew.
Clamps are designed for use in lattice systems, and fit rod sizes up to 3/4" diameter.
Customers Who Bought This Also Bought