SP Wilmad-LabGlass Metal Clamps, SP Industries
Supplier: Avantor
Certificates
About this item
These clamps hold Standard Taper joints in place securely under moderate pressure.
Made of metal, they are coated with silicone lacquer to avoid scratching glass surfaces. They can withstand temperatures up to 200°C, and are suitable where higher temperatures are used or cracking occurs in the plastic equivalent.
Customers Who Bought This Also Bought