ContactUsLinkComponent
Pressure Relief Valve, Chemglass
Pressure Relief Valve, Chemglass
Catalog # 89029-590
Supplier:  Chemglass
Small Business Enterprise
undefined
Pressure Relief Valve, Chemglass
Catalog # 89029-590
Supplier:  Chemglass
Supplier Number:  CG-999-01
Small Business Enterprise
Specifications
  • Description:
    Pressure Relief Valve, Complete
  • Cat. no.:
    89029-590
  • Supplier No.:
    CG-999-01
Specifications
About this item
Automatic, adjustable pressure relief valve is designed for use on an apparatus where internal pressure build-up is a concern
Tension on the stainless steel spring is adjusted by turning the top control knob in or out to relive pressures from 5 to 50 psi. Two-piece stem is made of PTFE and is fitted with Viton O-rings.