Specifications
- Volume:10 ml
- Description:Heparin/Glycerol Catheter Lock Solution
- Cat. no.:89134-258
- Supplier No.:HGS-10
Specifications
About this item
The antithrombotic characteristics of Heparin and the viscous nature of glycerol or dextrose are combined in a solution to promote and extend the patency of vascular catheters
- Used in both large and small species
- High osmolarity of dextrose prevents bacterial colonization
- High viscosity reduces washout from the catheter tip
- Effective physical barrier against clot formation
These solutions are proven performers in catheterized animal studies.