Specifications
- Description:QAC test kit
- Cat. no.:66170-492
- Range:0 - 500 ppm
- Supplier No.:3043-DR
- No. of Tests:50 (at 500 ppm)
Specifications
About this item
This kit tests for QAC using the direct reading titrator method and is best suited to higher QAC concentrations.
A masked bromphenol blue indicator is added to the sample and turns green
Range and sensitivity is 0 to 500 ppm/10 ppm alkyl dimethyl benzyl ammonium chloride.
Sodium tetraphenylboron is added to complex the QAC and the color changes to red.