To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Pfu DNA Ligase
- Size:400 unit
- Cat. no.:CA99902-306
Specifications
About this item
Pfu DNA ligase is a recombinant enzyme derived from Pyrococcus furiosus hyperthermophilic marine archaebacterium that catalyzes linkage of adjacent 5'-phosphate and 3'-hydroxy ends of double-stranded DNA at 45 to 80 °C, with a temperature optimum near 70 °C for nick-sealing reactions.
- Highly thermostable, with half-life of over 60 minutes at 95 °C
- Increases reliability of LCR by permitting higher melt temperatures
- Eliminates LCR template-predenaturation step, less enzyme required for LCR
It has higher ligation specificity and lower background than Tth DNA ligase.
Kit contains: 400 U Pfu DNA Ligase 10X reaction buffer