Specifications
- Description:AimTab™ Reducing Substances Tablets
- Cat. no.:76000-742
Specifications
About this item
Detects glucose, lactose, fructose, galactose, pentoses in urine, wine, or any other liquids.
- Replacement for Bayer's Clinitest tablet
- Test urine, stool, wine, or other liquids
- Designed for forensic use only
Tablet method for the semi-quantitative determination of the amount of reducing substances (glucose, lactose, fructose, galactose, pentoses) in urine, wine, or any other liquids. Compare to Clinitest, a trademark by Bayer Healthcare LLC.