To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Assay Type:Competitive
- Sensitivity:80% B/B0: 45 pg/mL
- Description:Kit contains: Antibody, Tracer, Standard, Buffers, Reagents, and Plate
- Mid point:50% B/B0: 150 – 250 pg/mL
- Detection range:15.6 - 2000 pg/ml
- Cat. no.:75816-668
- Supplier No.:10004023-96
Specifications
About this item
Fast, accurate measurement of TXB2.
- Measure TXB2 in multiple sample types
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure TXB2 levels down to 45 pg/ml
- Rapid assay; get results in under 4 hours
- Midpoint: 50% B/B0: 150-250 pg/ml
Thromboxane A2 (TXA2), produced from arachidonic acid, causes irreversible platelet aggregation and contraction of vascular and bronchial smooth muscle. TXA2 is rapidly hydrolyzed non-enzymatically to TXB2, which is then quickly metabolized to urinary metabolites for clearance by the kidneys. Urinary analysis of TXB2 accurately reflects intrarenal TXA2 synthesis, while measurement of 11-dehydro and 2,3-dinor TX metabolites gives the best estimate of systemic TXA2 secretion. Cayman’s TXB2 Express ELISA is a competitive assay that provides a rapid, accurate measurement of TXB2 from plasma, urine, and cell culture supernatants.