Uvex Hypershock™ Safety Eyewear, Honeywell Safety
Supplier: Honeywell Safety Products
Certificates
About this item
Uvex Hypershock™ saftey eyewear delivers the ultimate combination of high-performance protection and aggressive sport inspired styling.
- 100% dielectric
- Can be worn in environments where accidental exposure to electrical charge is possible
- Lightweight, wrap-around frame, side protection and superior peripheral vision
- Available in three frame colors and a wide variety of lens tints
- Polarized lenses in matte black and smoke brown frames
- Ideal for indoor and outdoor applications in construction, oil, gas and chemical, utilities, landscaping and manufacturing
Available with Uvextreme Plus® anti-fog lens coating for superior, long-lasting fog free vision that lasts up to 10X longer and provides 2X better scratch resistance than competitive anti-fog coated products. Also offered with scratch-resistant hardcoat lens coating for long-lasting protection from scuffing, scratching, and a variety of chemicals.
The full frame design offers high impact protection, with streamlined padded temples and a molded nosepiece for a secure, comfortable fit.
Certifications: Meets the ANSI Z87.1-2010 standard.
Customers Who Bought This Also Bought