PAL SPME Arrows and Kits, Restek
Supplier: Restek
Certificates
About this item
The PAL SPME Manual Injection Kit is designed to house SPME Arrows and traditional SPME fibers during extraction and injection steps.
- Rugged stainless steel construction
- Longer lifetime
- Faster extraction
- Higher sample throughput
- Better sensitivity allows lower LODs
Due to the relatively large diameter of Restek PAL SPME Arrows, you must modify the GC inlet using an instrument-specific conversion kit prior to use.
BodyTextAccessories: A GC inlet conversion kit is required in order to use a Restek PAL SPME Arrow.
Ordering information: These molecular weight ranges are a reasonable approximation; however, end users should verify suitability for their specific application. All Restek PAL SPME Arrows have 20 mm of phase bonded onto stainless steel.
Customers Who Bought This Also Bought