National Basic Sensor® Temperature Sensor with Dual Sensor Probe, Ace Glass
Supplier: Ace Glass
Certificates
About this item
Temperature sensor with two 72-inch SMP leads, for use with limit controllers, or for sending data to two different temperature devices simultaneously.
- Maximum sheath temperature: 550 °C
- 72-inch SMP leads
Inconel sheaths offer corrosion resistance and good high-temperature strength.
Customers Who Bought This Also Bought