Specifications
- Description:RS232 printer and computer cable set
- Cat. no.:10010-664
- Supplier No.:1010004
Specifications
About this item
This cable set can be used to connect a computer and printer to a benchtop or portable meter.
Designed for use with Orion™ Versa Star™ meters, Orion™ Star™ A series meters (Star A320, Star A220 and Star A210 series) and legacy Orion™ Star and Star Plus meters.