Specifications
- Description:RS-232 cable
- Cat. no.:14004-320
- Supplier No.:1010053
Specifications
About this item
Cable for connecting meters to a computer.
This cable is designed for use with Orion™ Versa Star meters, Orion™ Star™ A series meters (Star A320, Star A220 and Star A210 series) and legacy Orion™ Star™ and Star Plus meters.