Glycerine (synthetic) ≥95.0% FCC
Supplier: Avantor
Certificates
About this item
Synonyms: 1,2,3-Propanetriol,Propane-1,2,3-triol
Formula:HOCH₂CH(OH)CH₂OH
MW:92.09 g/mol
Density:1.264
MDL Number:MFCD00004722
CAS Number:56-81-5
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95,0
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 18.6 °C
:Flash Pt:
:Flash Pt:
:M.W. 92.09 g/mol
:18.6
:290
:177
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (C₃H₈O₃):95.0 - 100.5 %
- Specific Gravity @ 25°C:1.249
- Color:To Pass Test
- Chlorinated Compounds (As Cl):max. 0.003 %
- Fatty Acids and Esters (As Butyric Acid; Limit About 0.1 %):To Pass Test
- Glycerine (synthetic) ≥95.0% FCC:
Specification Test Results
Customers Who Bought This Also Bought