InSight® Ultra-Clear Bins, Akro-Mils
Supplier: Avantor
Certificates
About this item
A visible difference in storage and organization
- Unique "lock-in-place" design for secure stacking
- Strong molded hanger supports bins on hanging systems
- Lids protect contents from dust and damage
- Bins will stack with lids in place
- Made in the USA
These clear hang and stack bins are constructed of super-tough polycarbonate and are autoclavable for use in medical applications. Optional clear length and width dividers are available separately. InSight® Ultra-Clear bins are ideal for use in a variety of settings, from industrial to healthcare.
InSight® Ultra-Clear Bins provide easy and maximum visibility to users in a sleek, modern design.
BodyTextAccessories: Crystal clear dividers have fewer shadows and make parts easier to find.
Customers Who Bought This Also Bought