PIG® Grippy® Urinal Mat Runner, New Pig
Catalog #: 75982-610
Supplier: 2 D 2 S.A. DE C.V. MX
Specifications
- Description:PIG® Grippy® Urinal Mat Runner
- Cat. no.:75982-610
- Length:762 cm (25 ft.)
- Width:61 cm (2 ft.)
Specifications
About this item
Keep floors cleaner, drier, and safer with an adhesive-backed PIG® Grippy® Urinal Mat Runner for a more professional restroom environment.
- Industry-first design lets users roll out and cut to length the perfect size runner for continuous drip/splash coverage under a row of urinals
- Proprietary adhesive formulation applied across entire backing of mats ensures they stay in place with no shifting, curling, or flipping up
- Sticks to floors to stop slip and trip hazards, and eliminates time spent repositioning mats that have shifted
- Permanently infused with antimicrobial technology to prevent the growth of odor-causing bacteria and mold
- Top surface is engineered to absorb urine and water on contact, wick it away, and dry out quickly
- Durable, low-profile mats can be mopped without shifting or sliding
- Two impermeable backing layers ensure zero liquid pass-through, protecting floors and grout against degradation and discoloration from uric acid
- 25 foot roll provides enough coverage for up to 10 urinals; also ideal for rows of sinks or wherever custom sizes are needed
Customers Who Bought This Also Bought