PIG® Grippy® Toilet Floor Mat, New Pig
Catalog #: 75982-602
Supplier: New Pig
Specifications
- Description:PIG® Grippy® Toilet Mat
- Cat. no.:75982-602
- Length:39 cm (15.5")
- Width:46 cm (18")
Specifications
About this item
Absorb urine drips and splashes to keep floors cleaner, drier, and safer with adhesive-backed PIG® Grippy® Toilet Mats for a more professional restroom environment.
- Mat design fits around the toilet base to provide maximum drip/splash coverage
- Proprietary adhesive formulation applied across entire backing of mats ensures they stay in place with no shifting, curling, or flipping up
- Sticks to floors to stop slip and trip hazards, and eliminates time spent repositioning mats
- Permanently infused with antimicrobial technology to prevent the growth of odor-causing bacteria and mold
- Top surface is engineered to absorb urine and water on contact, wick it away, and dry out quickly
- Durable, low-profile mats can be mopped without shifting or sliding
- Two impermeable backing layers ensure zero liquid pass-through, protecting floors and grout against degradation and discoloration from uric acid
Customers Who Bought This Also Bought